Cassette Clamp, right - FAQs
Cassette Clamp, right - FAQs
View additional product information for Cassette Clamp, right - FAQs (B4478592)
1 product FAQs found
Can I purchase replacement parts for the Mini Blot Module?
Here are the replacement parts we offer for the Mini Blot Module:
- Mini Blot Module Gasket, Cat. No. B1001
- Gel Knife, Cat. No. EI9010
- Bolt Tank Base, Cat. No. B4478640
- Gel Runner Tank, Cat. No. B4478641
- Mini Gel Tank Lid, Cat. No. A25944
- Cassette Clamp, Right, Cat. No. B4478592
- Cassette Clamp, Left, Cat. No. B4478593
- Power Supply Adapters, Cat. No. ZA10001
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.