Choice™ Polypropylene (PP) Syringe Filters

Choice™ Polypropylene (PP) Syringe Filters

Use Choice Polypropylene Syringe Filters as an economic solution to filtration of biological samples, and for filtration of aggressive organic solutions.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)
CH2225-PP0.2 μm25 mm
CH4525-PP0.45 μm25 mm
CH2213-PP0.2 μm13 mm
CH4513-PP0.45 μm13 mm
Catalog number CH2225-PP
Price (USD)
96.12
Pack of 100
Pore Size:
0.2 μm
Diameter (Metric):
25 mm
Price (USD)
96.12
Pack of 100

Thermo Scientific™ Choice™ Polypropylene Syringe Filters are a good choice for filtration of biological samples, and for filtration of aggressive organic solutions. They provide cleaner sample extracts by removing interfering materials and fine particles down to the rated pore size.

Adding a syringe filtration step prior to chromatographic injection not only helps to ensure more consistent and reliable results, it also helps protect instruments and prolong column life. Economic Choice Syringe Filters are available in a variety of sizes and membrane types to meet any application need.

Specifications
  • Membrane: Hydrophilic polypropylene
  • Maximum operating temperature: 110 °C
  • Housing: Medical grade, virgin polypropylene
Applications
  • Protein- or peptide-based assays
  • General HPLC analysis
Chemical Incompatibilities
  • Hexane, toluene, benzene, limited resistance to dichloromethane and chloroform
Specifications
Diameter (Metric)25 mm
For Use With (Application)HPLC, SPE
Housing MaterialSolvent Resistent, Low-Extractable Polypropylene
PrefilterNo
Pressure Rating90 psi
Product LineChoice
Quantity100/Pk.
TypeSyringe Filter
DisposableSingle-use
MaterialPolypropylene
Material (Membrane)Polypropylene (Hydrophilic)
Pore Size0.2 μm
SterilityNon-sterile
Unit SizePack of 100