Human TNF alpha Matched Antibody Pair - FAQs
Human TNF alpha Matched Antibody Pair - FAQs
View additional product information for Human TNF alpha Matched Antibody Pair - FAQs (CHC1753)
1 product FAQs found
Does the Human TNF alpha ELISA Kit (Cat. No. KHC3011) use the same antibodies as TNF alpha Human Matched Antibody Pair (Cat. No. CHC1753)?
Yes, these kits use the same antibodies.