Endostatin Human ProcartaPlex™ Simplex Kit
Invitrogen™

Endostatin Human ProcartaPlex™ Simplex Kit

The Human Endostatin Simplex ProcartaPlex Kit measures endostatin protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.
Catalog NumberQuantity
EPX010-12238-90196 Tests
Catalog number EPX010-12238-901
Price (JPY)
41,200
Online offer
Ends: 25-Sep-2026
58,900
Save 17,700 (30%)
Each
Quantity:
96 Tests

The Human Endostatin Simplex ProcartaPlex Kit measures endostatin protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay RangeAs determined for Lot 1: 5 to 194,000 pg/mL
Assay SensitivityAs determined for Lot 1: 4.19 pg/mL
Bead TypeEndostatin [15]
For Use With (Equipment)Luminex™ Instruments
FormatSimplex Kit
GeneCollagen type XV alpha 1 chain
Gene ID (Entrez)P39059
Gene SymbolCOL15A1
Product LineProcartaPlex
ProteinEndostatin
Protein SubtypeCollagen alpha-1(XV) chain
Sample TypeSerum, Plasma, Cell Culture Supernatants
Sample VolumeSerum, Plasma: 25 μL, CCS: 50 μL
Shipping ConditionWet Ice
UniProt IDP39059
CombinabilityCombinable
Product TypeSimplex Kit
Quantity96 Tests
Research AreaImmunology, Oncology, Cardiovascular
SpeciesHuman
Unit SizeEach
Contents & Storage

• 1 vial Capture Beads (50X)
• 1 vial biotinylated Detection Antibody (50X)
• 2 vials Human Standard Mix V (lyophilized)
• 1 Bottle Universal Assay Buffer

Store at 2°C to 8°C.

Frequently asked questions (FAQs)

Do you offer a Mac-compatible version of the ProcartaPlex Analyst Software?

Unfortunately, we do not currently offer a Mac-compatible version of the ProcartaPlex Analyst Software. ProcartaPlex Analyst Software is compatible with PC, or xPONENT Software is available on Luminex instruments to analyze your data.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am getting poor recovery with my ProcartaPlex multiplex immunoassay. What can I do?

Here are possible causes and solutions:
- The wrong cell culture media may have been used to prepare the standards. We recommend using the same cell culture media that was used to culture the cells.
- The samples and antigen standards may not have been stored on ice. We recommend preparing the samples and standards on ice before setting up the assay.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am seeing low signal or sensitivity in my ProcartaPlex multiplex immunoassay. What should I do?

Standards may not have been reconstituted and diluted correctly. We recommend preparing fresh antigen standards following the instructions provided in the corresponding ProcartaPlex user guide.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

With my ProcartaPlex multiplex immunoassay, I am getting a low bead count. What is causing this?

Here are possible causes and solutions:
- Volume of the bead solution is too low. We recommend adding 120 µL Reading Buffer into each well and shaking at 600 rpm for 5 mins at room temperature to resuspend beads before reading on the Luminex instrument.
- High bead aggregation. We recommend vortexing the bead suspension well before using in the assay and ensuring that the beads are properly mixed during the incubation steps.
- Dyes contained in the beads are photo-bleached from overexposure to light. We recommend storing the bead solution and the 96-well plate in the dark.
- Samples causing the instrument to clog. We recommend removing the 96-well Flat Bottom Plate and performing a wash and rinse to the instrument and then re-running the assay with further dilution of samples.
- Probe height is incorrect. We recommend referring to the Luminex manual for proper adjustment of the needle height.
- The instrument needle is partially clogged. We recommend replacing or cleaning the needle according to the manufacturer's recommendations.
- Beads stuck to the bottom of the plate. We recommend confirming that the plate shaker is set to 600 rpm and shaking for at least 5 mins before reading.
- Air bubble in the sample loop. We recommend referring to the specific Luminex manual for your assay for proper removal of the air bubble.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

In ProcartaPlex multiplex immunoassays, we are seeing a limited dynamic range for BioPlex software users. Why is this?

The instrument may have been calibrated at high PMT settings. We recommend calibrating the instrument using the CAL2 Low RP1 target value.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.