Revco™ ExF -86°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capacity, holds 240 boxes, 120V/60Hz, 16A, painted galvaneel interior - FAQs
View additional product information for Revco™ ExF -86°C Upright Ultra-Low Temperature Freezers - FAQs (EXF24086A)