FluoSpheres™ Carboxylate-Modified Microspheres
Invitrogen™

FluoSpheres™ Carboxylate-Modified Microspheres

Achieve the brightest fluorescence with Carboxylate-Modified FluoSphere Microspheres, available in different colors and particle sizes.
Catalog NumberDiameter (Metric)ColorExcitation/EmissionQuantity
F87930.04 μmRed580/605 nm1 mL
F88030.1 μmYellow-Green505/515 nm10 mL
F88161.0 μmCrimson625/645 nm2 mL
F88231.0 μmYellow-Green505/515 nm10 mL
F88010.1 μmRed580/605 nm10 mL
F88110.2 μmYellow-Green505/515 nm10 mL
F88070.2 μmDark Red660/680 nm2 mL
F107200.04 μmYellow-Green, Orange, Red, Dark Red505/515, 540/560, 580/605, 660/680 nm1 mL/each
F208810.2 μmOrange365/610 nm2 mL
F8783
also known as F-8783
0.02 μmDark Red660/680 nm2 mL
F87860.02 μmRed580/605 nm10 mL
F8795
also known as F-8795
0.04 μmYellow-Green505/515 nm1 mL
F88130.5 μmYellow-Green505/515 nm10 mL
F88201.0 μmOrange540/560 nm10 mL
F88252.0 μmNile Red535/575 nm2 mL
F88272.0 μmYellow-Green505/515 nm2 mL
F8781
also known as F-8781
0.02 μmBlue365/415 nm10 mL
F8782
also known as F-8782
0.02 μmCrimson625/645 nm2 mL
F87840.02 μmNile Red535/575 nm10 mL
F87870.02 μmYellow-Green505/515 nm10 mL
F8789
also known as F-8789
0.04 μmDark Red660/680 nm1 mL
F8792
also known as F-8792
0.04 μmOrange540/560 nm1 mL
F87940.04 μmRed-Orange565/580 nm1 mL
F87970.1 μmBlue350/440 nm10 mL
F87990.1 μmInfrared715/755 nm1 mL
F8800
also known as F-8800
0.1 μmOrange540/560 nm10 mL
F88050.2 μmBlue365/415 nm10 mL
F88060.2 μmCrimson625/645 nm2 mL
F88090.2 μmOrange540/560 nm10 mL
F88100.2 μmRed580/605 nm10 mL
F88120.5 μmRed580/605 nm10 mL
F88141.0 μmBlue365/415 nm10 mL
F88151.0 μmBlue350/440 nm10 mL
F88191.0 μmNile Red535/575 nm10 mL
F88211.0 μmRed580/605 nm10 mL
F8824
also known as F-8824
2.0 μmBlue365/415 nm2 mL
F88262.0 μmRed580/605 nm2 mL
Catalog number F8793
Price (JPY)
98,800
Each
Diameter (Metric):
0.04 μm
Color:
Red
Excitation/Emission:
580/605 nm
Quantity:
1 mL

Easily perform flow cytometry, microscopy, HTS, HCS, immunoassay, and other laboratory applications using our extensive selection of FluoSpheres Carboxylate-Modified Microspheres. FluoSphere beads can be used in passive adsorption or active, covalent coupling of proteins, nucleic acids, and biomolecules for particle capture applications. FluoSphere microspheres are loaded with proprietary fluorescent dyes, making them the brightest microspheres available.

Visualize the brightest fluorescence for laboratoy applications including fluorescence microscopy, flow cytometry, HTS, HCS, and cell tracing with our Carboxylate-Modified FluoSphere Microspheres, which are manufactured from polystyrene microspheres and loaded with different proprietary dyes. Using specialized staining methods enables all of the fluorescent dye molecules to be contained inside each polystyrene microsphere instead of on the bead's surface. This protective environment within the bead shields the dye from detrimental environmental effects, such as photobleaching. Our carboxylate-modified microspheres are coated with a hydrophilic polymer containing multiple carboxylic acids for covalent attachment of ligands. A range of particle sizes is available for different research uses and experiments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation/Emission580/605 nm
Product LineFLUOSPHERES
Quantity1 mL
Surface ModificationCarboxylate
ColorRed
Diameter (Metric)0.04 μm
For Use With (Application)Fluorescence Microscopy
MaterialPolystyrene
Product TypeCarboxylate-Modified Microsphere
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

I read that carboxylate-modified latex (CML) beads have a "fluffy coat" of carboxyl groups on their surface. What is meant by this?

The CML beads have a high density of carboxyl groups at the surface. The surface layer is quite hydrophilic and at the appropriate pH (pH>5), are charged; due to electrostatic repulsion, this type of surface is 3-dimensional and may be considered analogous to the fuzz on a tennis ball.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I have some FluoSpheres polystyrene microspheres, with 20 nm diameter. They are aggregating a lot. What can I do about it?

The smaller the microspheres, the greater the propensity to aggregate. But the aggregation is not irreversible. Sonicate in a bath sonicator or vortex to disperse, just prior to use. You can also add a small concentration of Tween-20 or Triton X-100 (unless you are using them in a live-cell system).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I sonicated my 2.0 µm carboxylate-modified microspheres, as recommended, but saw foaming (bubbles) on top of the solution. Should I be concerned?

Use of a bath sonicator is recommended to help break up any aggregated microspheres. The foaming is from Tween-20, which is in the stock solution to help prevent aggregation. It is normal and expected to see bubbles from this. Do not use a probe sonicator, which would cause damage to the microspheres (as well as much more bubbling).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the warranty for FluoSpheres microspheres?

The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After washing and centrifugation, there was only a very small pellet left of my microsphere beads and the solution was transparent. Why is this?

Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:

Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (9)

Citations & References
Abstract
Distribution of retrogradely transported fluorescent latex microspheres in rat lumbosacral ventral root axons following peripheral crush injury: a light and electron microscopic study.
Authors:Persson HG, Gatzinsky KP
Journal:Brain Res
PubMed ID:8118679
'The retrograde axonal transport of fluorescent latex microspheres, which are tracers extensively used for studying the neuronal connectivity in the CNS, was investigated in large myelinated lumbosacral ventral root nerve fibres of adult rats following peripheral crush injury. After crushing the sciatic nerve, a suspension of 30 nm red-fluorescent latex ... More
Identification of catecholaminergic inputs to and outputs from aromatase-containing brain areas of the Japanese quail by tract tracing combined with tyrosine hydroxylase immunocytochemistry.
Authors:Balthazart J, Absil P
Journal:J Comp Neurol
PubMed ID:9183702
'In the quail brain, aromatase-immunoreactive (ARO-ir) neurons located in the medial preoptic nucleus (POM) and caudal paleostriatum ventrale/nucleus accumbens/nucleus striae terminalis complex (PVT/nAc/nST) receive catecholaminergic inputs identified by the presence of tyrosine hydroxylase-immunoreactive (TH-ir) fibers and punctate structures. The origin of these inputs was analyzed by retrograde tracing with cholera ... More
The origins of catecholaminergic innervation in the rostral ventromedial medulla oblongata of the rat.
Authors:Tanaka M, Matsumoto Y, Murakami T, Hisa Y, Ibata Y
Journal:Neurosci Lett
PubMed ID:8710209
'The localization of catecholaminergic neuronal cell bodies which project to the rostral ventromedial medulla oblongata (RVM) were investigated by the combined technique with dopamine beta-hydroxylase (DBH), phenylethanolamine N-methyltransferase (PNMT) or tyrosine hydroxylase (TH) immunocytochemistry and retrograde neuronal tracing method using fluorescent latex microspheres (FluoSpheres) injected into the center of the ... More
Optimized localization analysis for single-molecule tracking and super-resolution microscopy.
Authors:Mortensen KI, Churchman LS, Spudich JA, Flyvbjerg H,
Journal:Nat Methods
PubMed ID:20364147
'We optimally localized isolated fluorescent beads and molecules imaged as diffraction-limited spots, determined the orientation of molecules and present reliable formulas for the precision of various localization methods. Both theory and experimental data showed that unweighted least-squares fitting of a Gaussian squanders one-third of the available information, a popular formula ... More
T-type channels control the opioidergic descending analgesia at the low threshold-spiking GABAergic neurons in the periaqueductal gray.
Authors:Park C, Kim JH, Yoon BE, Choi EJ, Lee CJ, Shin HS,
Journal:Proc Natl Acad Sci U S A
PubMed ID:20682748
'Endogenous opioids generate analgesic signals in the periaqueductal gray (PAG). However, because cell types in the PAG are difficult to identify, its neuronal mechanism has remained poorly understood. To address this issue, we characterized PAG neurons by their electrical properties using differentially labeled GABAergic and output neurons in the PAG. ... More