Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Fisher Chemical™

Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™

ClH, CAS Number-7647-01-0, muriatic acid, spirits of salt, hydrogen chloride hcl, hydrogen chloride, hydrochloric acid, chlorwasserstoff, chloorwaterstof, chlorohydric acid, acide chlorhydrique, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol | CAS: 7647-01-0 | ClH | 36.46 g/mol
Quantity:
1 L
2.5 L
Packaging:
View all
HDPE plastic bottle
Catalog number H-1200-PB15
also known as H/1200/PB15
Price (JPY)
-
Contact Us ›
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Boiling Point57°C
CAS7647-01-0
ChEBICHEBI:17883
ColorColorless
Formula Weight36.46g/mol
View more
ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, chlorwasserstoff, chloorwaterstof, muriatic acid, acide chlorhydrique, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol