4-Fluoropiperidine hydrochloride, 95%
4-Fluoropiperidine hydrochloride, 95%
4-Fluoropiperidine hydrochloride, 95%
4-Fluoropiperidine hydrochloride, 95%
Thermo Scientific Chemicals

4-Fluoropiperidine hydrochloride, 95%

CAS: 57395-89-8 | C5H11ClFN | 139.598 g/mol
Quantity:
250 mg
1 g
Catalog number H25755.03
also known as H25755-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS57395-89-8
IUPAC Name4-fluoropiperidine hydrochloride
Molecular FormulaC5H11ClFN
InChI KeyIXENWFQXVCOHAZ-UHFFFAOYSA-N
SMILESCl.FC1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Water Content (Karl Fischer Titration)≤3.0%
Melting Point (clear melt)152.0-169.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only