(1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate
(1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate
(1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate
Thermo Scientific Chemicals

(1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate

CAS: 12245-39-5 | C13H19O2Rh | 310.20 g/mol
Quantity:
250 mg
1 g
Catalog number H26083.MD
also known as H26083-MD
Price (JPY)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS12245-39-5
IUPAC Nameλ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2Z)-4-oxopent-2-en-2-olate
Molecular FormulaC13H19O2Rh
InChI KeyTVGPAHONTUMCNP-DWVXZKBMSA-M
SMILES[Rh+].C\C([O-])=C\C(C)=O.C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormCrystalline powder
Assay (special)≥98.6 to ≤101.6% by rhodium elemental analysis
Elemental AnalysisRhodium, Rh: 32.7-33.7%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only