3-Amino-5-(trifluoromethyl)benzoic acid, 97%
3-Amino-5-(trifluoromethyl)benzoic acid, 97%
3-Amino-5-(trifluoromethyl)benzoic acid, 97%
Thermo Scientific Chemicals

3-Amino-5-(trifluoromethyl)benzoic acid, 97%

CAS: 328-68-7 | C8H6F3NO2 | 205.14 g/mol
Quantity:
1 g
5 g
Catalog number H26169.03
also known as H26169-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS328-68-7
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥96.0%
Appearance (Color)Pale cream to cream
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)138.0-148.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only