2,6-Dimethyl-4-nitroanisole, 99%
2,6-Dimethyl-4-nitroanisole, 99%
2,6-Dimethyl-4-nitroanisole, 99%
2,6-Dimethyl-4-nitroanisole, 99%
Thermo Scientific Chemicals

2,6-Dimethyl-4-nitroanisole, 99%

CAS: 14804-39-8 | C9H11NO3 | 181.19 g/mol
Quantity:
1 g
5 g
Catalog number H26204.03
also known as H26204-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS14804-39-8
IUPAC Name2-methoxy-1,3-dimethyl-5-nitrobenzene
Molecular FormulaC9H11NO3
InChI KeyHSDNHFOJTRMGER-UHFFFAOYSA-N
SMILESCOC1=C(C)C=C(C=C1C)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
CommentYellow
Melting Point (clear melt)87.0-93.0?C
Assay (GC)≥98.5%
Water Content (Karl Fischer Titration)≤1.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only