4-(Trimethylsilyl)diphenylacetylene, 99%
4-(Trimethylsilyl)diphenylacetylene, 99%
4-(Trimethylsilyl)diphenylacetylene, 99%
Thermo Scientific Chemicals

4-(Trimethylsilyl)diphenylacetylene, 99%

CAS: 136459-72-8 | C17H18Si | 250.416 g/mol
Quantity:
1 g
5 g
25 g
Catalog number H26300.14
also known as H26300-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS136459-72-8
IUPAC Nametrimethyl[4-(2-phenylethynyl)phenyl]silane
Molecular FormulaC17H18Si
InChI KeyUZRJWRFLYPENFH-UHFFFAOYSA-N
SMILESC[Si](C)(C)C1=CC=C(C=C1)C#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.5%
Appearance (Color)White to pale cream
Melting Point (clear melt)35.0-42.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only