Di-tert-butylphenylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals
Di-tert-butylphenylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals
Di-tert-butylphenylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-tert-butylphenylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog NumberQuantity
H26904.03
also known as H26904-03
1 g
Catalog number H26904.03
also known as H26904-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS612088-55-8
IUPAC Namedi-tert-butyl(phenyl)phosphanium; tetrafluoroboranuide
Molecular FormulaC14H24BF4P
InChI KeyHRDPEVWZXUWEFR-UHFFFAOYSA-O
SMILESF[B-](F)(F)F.CC(C)(C)[PH+](C1=CC=CC=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Phosphorus-31 NMRConforms to structure
FormCrystalline powder
Proton NMRConforms to structure
Assay by NMR≥98.5% by Phosphorus-31 NMR
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only