N-Benzyloxycarbonyl-L-alaninol is used as an important organic intermediate. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzyloxycarbonyl-L-alaninol is used as an important organic intermediate. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only
General References:
- K Hanabusa.; K Okui.; K Karaki.; T Koyama. A small molecular gelling agent for organic liquids: N-benzyloxycarbonyl-L-alanine 4-hexadecanoyl-2-nitrophenyl ester. J. Chem. Soc., Chem. 19921 (18), 1371-1373.
- YZ An.; JL Anderson.; Y Rubin. Synthesis of. alpha.-amino acid derivatives of C60 from 1, 9-(4-hydroxycyclohexano) buckminsterfullerene. J. Org. Chem. 199358 (18), 4799-4801.