3-(1H-Tetrazol-5-yl)phenol, 97%
3-(1H-Tetrazol-5-yl)phenol, 97%
3-(1H-Tetrazol-5-yl)phenol, 97%
3-(1H-Tetrazol-5-yl)phenol, 97%
Thermo Scientific Chemicals

3-(1H-Tetrazol-5-yl)phenol, 97%

CAS: 96859-34-6 | C7H6N4O | 162.15 g/mol
Quantity:
250 mg
1 g
5 g
Catalog number H27529.03
also known as H27529-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS96859-34-6
IUPAC Name3-(2H-1,2,3,4-tetrazol-5-yl)phenol
Molecular FormulaC7H6N4O
InChI KeyIZORRBUQWFSCII-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)C1=NNN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only