4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals
4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals
4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog NumberQuantity
H27562.14
also known as H27562-14
25 g
Catalog number H27562.14
also known as H27562-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS55685-75-1
IUPAC Name(4-chloro-2-methoxyphenyl)methanol
Molecular FormulaC8H9ClO2
InChI KeyAXXPZAITCCIOIA-UHFFFAOYSA-N
SMILESCOC1=C(CO)C=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)44.0-54.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only