2-Cyclohexylethylamine, 97%
2-Cyclohexylethylamine, 97%
2-Cyclohexylethylamine, 97%
Thermo Scientific Chemicals

2-Cyclohexylethylamine, 97%

CAS: 4442-85-7 | C8H17N | 127.23 g/mol
Quantity:
1 g
5 g
Catalog number H27987.06
also known as H27987-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS4442-85-7
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4620-1.4670 @ 20?C
In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Protect from direct light.
RUO – Research Use Only

General References:

  1. Bingzhi Shi,; Tanjore K Narayanan,; Zhi-Ying Yang,; Bradley T Christian,; Jogeshwar Mukherjee. Radiosynthesis and in vitro evaluation of 2-(N-alkyl-N-1'-11C-propyl)amino-5-hydroxytetralin analogs as high affinity agonists for dopamine D-2 receptors. Nuclear Medicine and Biology. 1999, 26 (7),725-735.
  2. Beverly A Clement,; Christina M Goff,; T.David A Forbes. Toxic amines and alkaloids from acacia rigidula Phytochemistry. 1998, 49 (5),1377-1380.
  3. Kenichi Sakaia,; Rumiko Sakuraia,; Noriaki Hirayama. Molecular mechanism of DCR phenomenon observed in (RS)-1-cyclohexylethylamine-mandelic acid resolution system. Tetrahedron: Asymmetry. 2006, 17 (12),1812-1816.