Ethyl 7-bromoheptanoate, 97%
Ethyl 7-bromoheptanoate, 97%
Ethyl 7-bromoheptanoate, 97%
Thermo Scientific Chemicals

Ethyl 7-bromoheptanoate, 97%

CAS: 29823-18-5 | C9H17BrO2 | 237.14 g/mol
Catalog NumberQuantity
H28001.AA
also known as H28001-AA
5 mL
Catalog number H28001.AA
also known as H28001-AA
Price (JPY)
-
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
CAS29823-18-5
IUPAC Nameethyl 7-bromoheptanoate
Molecular FormulaC9H17BrO2
InChI KeyOOBFNDGMAGSNKA-UHFFFAOYSA-N
SMILESCCOC(=O)CCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4570-1.4620 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only