3-Bromo-4-methylphenol, 98%
3-Bromo-4-methylphenol, 98%
3-Bromo-4-methylphenol, 98%
3-Bromo-4-methylphenol, 98%
Thermo Scientific Chemicals

3-Bromo-4-methylphenol, 98%

CAS: 60710-39-6 | C7H7BrO | 187.036 g/mol
Quantity:
1 g
5 g
Catalog number H28689.03
also known as H28689-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS60710-39-6
IUPAC Name3-bromo-4-methylphenol
Molecular FormulaC7H7BrO
InChI KeyGMZKNRDHSHYMHG-UHFFFAOYSA-N
SMILESCC1=C(Br)C=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥97.5%
Appearance (Color)White to cream
Melting Point (clear melt)52.0-58.0?C
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. L.Haskelberg. The halogenation of aryloxyacetic acids and their homologs. J. Org. Chem. 1947, 12(3), 426-433.
  2. Dipl.-Chem. Michael Prein; Michael Maurer; Eva Maria Peters; Dr. Karl Peters; Prof. Dr. Hans Georg von Schnering; Prof. Dr. Waldemar Adam. Diastereoselective Synthesis of 4-Hydroperoxy-3,5-cyclohexadienones in the Photooxygenation of Hydroxyethyl-Substituted Phenols.Chemistry - A European Journal. 1995, 1(1), 89-94.