3-Methyl-5-nitropyridine, 97%, Thermo Scientific Chemicals
3-Methyl-5-nitropyridine, 97%, Thermo Scientific Chemicals
3-Methyl-5-nitropyridine, 97%, Thermo Scientific Chemicals
3-Methyl-5-nitropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-5-nitropyridine, 97%, Thermo Scientific Chemicals

Catalog NumberQuantity
H31683.06
also known as H31683-06
5 g
Catalog number H31683.06
also known as H31683-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6960-20-9
IUPAC Name3-methyl-5-nitropyridine
Molecular FormulaC6H6N2O2
InChI KeyOQWXZZCTJZOSGH-UHFFFAOYSA-N
SMILESCC1=CN=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)92.5-98.5?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)Pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only