2,3-Dibromopyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
2,3-Dibromopyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
2,3-Dibromopyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dibromopyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog NumberQuantity
H31708.03
also known as H31708-03
1 g
Catalog number H31708.03
also known as H31708-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1020056-98-7
IUPAC Name2,3-dibromopyridine-4-carboxylic acid
Molecular FormulaC6H3Br2NO2
InChI KeyICISUZSRQHUJKX-UHFFFAOYSA-N
SMILESOC(=O)C1=C(Br)C(Br)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥96.0%
Appearance (Color)Pale brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only