6-Bromochromone-3-carboxylic acid, 97%
6-Bromochromone-3-carboxylic acid, 97%
6-Bromochromone-3-carboxylic acid, 97%
Thermo Scientific Chemicals

6-Bromochromone-3-carboxylic acid, 97%

CAS: 51085-91-7 | C10H5BrO4 | 269.05 g/mol
Quantity:
1 g
5 g
Catalog number H31905.03
also known as H31905-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS51085-91-7
IUPAC Name6-bromo-4-oxo-4H-chromene-3-carboxylic acid
Molecular FormulaC10H5BrO4
InChI KeyIJTWNMVKNJSGMS-UHFFFAOYSA-N
SMILESOC(=O)C1=COC2=CC=C(Br)C=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Silylated GC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only