3-Bromo-4-heptanone, 98%
3-Bromo-4-heptanone, 98%
3-Bromo-4-heptanone, 98%
3-Bromo-4-heptanone, 98%
Thermo Scientific Chemicals

3-Bromo-4-heptanone, 98%

CAS: 42330-10-9 | C7H13BrO | 193.08 g/mol
Quantity:
1 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H32094.03
also known as H32094-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS42330-10-9
IUPAC Name3-bromoheptan-4-one
Molecular FormulaC7H13BrO
InChI KeySFKVBRLKXVRUQW-UHFFFAOYNA-N
SMILESCCCC(=O)C(Br)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4580-1.4620 @ 20?C
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only