Methyl 4-amino-3-bromo-5-nitrobenzoate, 97%, Thermo Scientific Chemicals
Methyl 4-amino-3-bromo-5-nitrobenzoate, 97%, Thermo Scientific Chemicals
Methyl 4-amino-3-bromo-5-nitrobenzoate, 97%, Thermo Scientific Chemicals
Methyl 4-amino-3-bromo-5-nitrobenzoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-amino-3-bromo-5-nitrobenzoate, 97%, Thermo Scientific Chemicals

Catalog NumberQuantity
H33216.06
also known as H33216-06
5 g
Catalog number H33216.06
also known as H33216-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS105655-17-2
IUPAC Namemethyl 4-amino-3-bromo-5-nitrobenzoate
Molecular FormulaC8H7BrN2O4
InChI KeyPRTLMEWWYBJZPN-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=C(N)C(Br)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)103.0-113.0?C
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only