4'-Chlorobiphenyl-4-sulfonyl chloride, 96%
4'-Chlorobiphenyl-4-sulfonyl chloride, 96%
4'-Chlorobiphenyl-4-sulfonyl chloride, 96%
4'-Chlorobiphenyl-4-sulfonyl chloride, 96%
Thermo Scientific Chemicals

4'-Chlorobiphenyl-4-sulfonyl chloride, 96%

CAS: 20443-74-7 | C12H8Cl2O2S | 287.154 g/mol
Quantity:
1 g
10 g
Catalog number H34182.03
also known as H34182-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS20443-74-7
IUPAC Name4'-chloro-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular FormulaC12H8Cl2O2S
InChI KeyNWYUSJMIHFIMTA-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
Melting Point (clear melt)106.0-115.0?C
Appearance (Color)White
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only