3-(3-Methylphenyl)propionic acid, 96%
3-(3-Methylphenyl)propionic acid, 96%
3-(3-Methylphenyl)propionic acid, 96%
3-(3-Methylphenyl)propionic acid, 96%
Thermo Scientific Chemicals

3-(3-Methylphenyl)propionic acid, 96%

CAS: 3751-48-2 | C10H12O2 | 164.204 g/mol
Quantity:
1 g
5 g
Catalog number H34395.06
also known as H34395-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3751-48-2
IUPAC Name3-(3-methylphenyl)propanoic acid
Molecular FormulaC10H12O2
InChI KeyVWXVTHDQAOAENP-UHFFFAOYSA-N
SMILESCC1=CC=CC(CCC(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (Silylated GC)≥95.0%
Melting Point (clear melt)37.5-46.5°C
FormCrystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only