4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
Thermo Scientific Chemicals

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%

CAS: 128852-17-5 | C8H9ClO2S2 | 236.73 g/mol
Quantity:
250 mg
1 g
Catalog number H34470.03
also known as H34470-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS128852-17-5
IUPAC Name4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride
Molecular FormulaC8H9ClO2S2
InChI KeyUUBSHLWNADKCBU-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC2=C(CCCC2)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0%
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Melting Point (clear melt)57.5-69.5?C
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only