You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
Ethylboronic acid pinacol ester, 99%
Element Search
Structure Search
Thermo Scientific™
Ethylboronic acid pinacol ester, 99%
CAS: 82954-89-0 | C8H17BO2 | 156.03 g/mol
Technical Support
Customer Service
Change view
Quantity:
1 g
5 g
25 g
Try the chem dex
TM
web app
Catalog number
H36868.14
also known as H36868-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS
82954-89-0
IUPAC Name
2-ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Formula
C8H17BO2
InChI Key
NPUBDPDASOEIOA-UHFFFAOYSA-N
SMILES
CCB1OC(C)(C)C(C)(C)O1
View more
Specifications
Specification Sheet
Assay (GC)
>98.5%
Proton NMR
Conform to structure
Form
Liquid
Appearance (Color)
Clear colorless