Specifications
CAS87394-55-6
Chemical Name or Material1-(3-Chloro-2-pyridyl)piperazine
Formula Weight197.67
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more