Specifications
CAS53712-75-7
Chemical Name or Material4-Cyclopropyl-4-oxobutyric acid
Formula Weight142.15
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more