2-Iodo-1-methylimidazole, 97%
2-Iodo-1-methylimidazole, 97%
2-Iodo-1-methylimidazole, 97%
Thermo Scientific Chemicals

2-Iodo-1-methylimidazole, 97%

CAS: 37067-95-1 | C4H5IN2 | 208.002 g/mol
Quantity:
250 mg
1 g
Catalog number H51862.03
also known as H51862-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS37067-95-1
Chemical Name or Material2-Iodo-1-methylimidazole
Formula Weight208
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only