2-(Diphenylphosphino)benzaldehyde oxime, 95%, Thermo Scientific Chemicals
2-(Diphenylphosphino)benzaldehyde oxime, 95%, Thermo Scientific Chemicals
2-(Diphenylphosphino)benzaldehyde oxime, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Diphenylphosphino)benzaldehyde oxime, 95%, Thermo Scientific Chemicals

Catalog NumberQuantity
H51903.06
also known as H51903-06
5 g
Catalog number H51903.06
also known as H51903-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS153358-05-5
Chemical Name or Material2-(Diphenylphosphino)benzaldehyde oxime
Formula Weight305.32
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Lucía Menéndez-Rodríguez, et al. Palladium(II) complexes with a phosphino-oxime ligand: synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.Catal. Sci. Technol.,2015,5(7), 3754-3761.