9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%
Thermo Scientific Chemicals

9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%

CAS: 196207-58-6 | C41H64B2O4 | 642.579 g/mol
Quantity:
1 g
5 g
Catalog number H51955.03
also known as H51955-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS196207-58-6
IUPAC Name2-[9,9-dioctyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC41H64B2O4
InChI KeyFAHIZHKRQQNPLC-UHFFFAOYSA-N
SMILESCCCCCCCCC1(CCCCCCCC)C2=C(C=CC(=C2)B2OC(C)(C)C(C)(C)O2)C2=C1C=C(C=C2)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥94.0% (HPLC)
Appearance (Color)White
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester is a reactant in the synthesis of 6,6-(5,5-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis(thiophene-5,2-diyl))bis(2,5-bis(2-ethylhexyl)-3-(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione) (DPP1), a solution-processable nonfullerene electron acceptor used in organic solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester is a reactant in the synthesis of 6,6-(5,5-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis(thiophene-5,2-diyl))bis(2,5-bis(2-ethylhexyl)-3-(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione) (DPP1), a solution-processable nonfullerene electron acceptor used in organic solar cells.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Alessandra Sergent; Gael Zucchi; Robert B.Pansu; Marc Chaigneau; Bernard Geffroy; Denis Tondelier; Michel Ephritikhine. Synthesis, characterization, morphological behaviour, and photo- and electroluminescence of highly blue-emitting fluorene-carbazolecopolymers with alkyl side-chains of different lengths. J. Mater. Chem. C. 2013, 1, (19),3207-3216
  2. Xiaoguang Huang; Gael Zucchi; Jacqueline Tran; Robert B.Pansu; Arnaud Brosseau; Bernard Geffroy; Francois Nief. Visible-emitting hybrid sol-gel materials comprising lanthanide ions: thin film behaviour and potential use as phosphors for solid-state lighting. New J. Chem. 2014, 38, (12), 5793-5800