3-(3-Benzothienyl)-N-Fmoc-L-alanine, 95%
3-(3-Benzothienyl)-N-Fmoc-L-alanine, 95%
3-(3-Benzothienyl)-N-Fmoc-L-alanine, 95%
Thermo Scientific Chemicals

3-(3-Benzothienyl)-N-Fmoc-L-alanine, 95%

CAS: 177966-60-8 | C26H21NO4S | 443.52 g/mol
Quantity:
250 mg
1 g
5 g
Catalog number H52054.MD
also known as H52054-MD
Price (JPY)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS177966-60-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormFused solid
Assay (HPLC)≥94.0%
Assay from Supplier's CofAD-Enantiomer ≤ 0.5%
Assay from Supplier's CofAMass Spec conforms
View more
It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility (second only to glycine) when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility (second only to glycine) when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only