N-Benzoyl-2'-deoxycytidine, 98+%
N-Benzoyl-2'-deoxycytidine, 98+%
N-Benzoyl-2'-deoxycytidine, 98+%
N-Benzoyl-2'-deoxycytidine, 98+%
Thermo Scientific Chemicals

N-Benzoyl-2'-deoxycytidine, 98+%

CAS: 4836-13-9 | C16H17N3O5 | 331.33 g/mol
Quantity:
1 g
5 g
Catalog number H52298.03
also known as H52298-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS4836-13-9
IUPAC NameN-{1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
Molecular FormulaC16H17N3O5
InChI KeyMPSJHJFNKMUKCN-OUCADQQQSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (HPLC)≥98.0%
Proton NMRConforms to structure
Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine, N-Protected 2?-Deoxyribonucleosides(useful for oligonucleotide synthesis), 3?-, 5?-and 3?, 5?-di-O-crotonyl 2?-deoxynucleoside derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only