N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%
Thermo Scientific Chemicals

N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%

CAS: 51549-39-4 | C23H31N5O4Si | 469.62 g/mol
Quantity:
1 g
5 g
Catalog number H52309.06
also known as H52309-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS51549-39-4
Chemical Name or MaterialN-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine
Formula Weight469.62
IUPAC NameN-{9-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide
InChI KeyBJZZQJFYKQPUGO-TWAUYLRJNA-N
View more
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoyl-5′-O-tert-butyldimethylsilyl-2′-deoxyadenosine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. S.K.Burley; A.H.J.Wang. Structure of N-benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine monohydrate. Acta Cryst. 1987, C43, 988-990