Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Chi B. Vu,; Deborah Pan,; Bo Peng,; Gnanasambandam Kumaravel,; Glenn Smits,; Xiaowei Jin,; Deepali Phadke,; Thomas Engber,; Carol Huang,; Jennifer Reilly,; Stacy Tam,; Donna Grant , Gregg Hetu,;Russell C. Petter. Novel Diamino Derivatives of [1,2,4]Triazolo[1,5-a][1,3,5]triazine as Potent and Selective Adenosine A2a Receptor Antagonists . J. Med. Chem.,. 2005, 48 (6),2009-2018.