trans-3-Heptene, 97%
trans-3-Heptene, 97%
trans-3-Heptene, 97%
Thermo Scientific Chemicals

trans-3-Heptene, 97%

CAS: 14686-14-7 | C7H14 | 98.19 g/mol
Quantity:
1 g
5 g
25 g
Catalog number H53482.03
also known as H53482-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS14686-14-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay from Suppliers CofA≥96.0% (GC)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only