2-Amino-4-(4-biphenylyl)thiazole, 97%
2-Amino-4-(4-biphenylyl)thiazole, 97%
2-Amino-4-(4-biphenylyl)thiazole, 97%
2-Amino-4-(4-biphenylyl)thiazole, 97%
Thermo Scientific Chemicals

2-Amino-4-(4-biphenylyl)thiazole, 97%

CAS: 2834-79-9 | C15H12N2S | 252.335 g/mol
Quantity:
250 mg
1 g
5 g
Catalog number H54413.03
also known as H54413-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein187642
CAS2834-79-9
Chemical Name or Material2-Amino-4-(4-biphenylyl)thiazole
Formula Weight252.33
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only