2-Fluoro-N-methylbenzylamine, 95%
2-Fluoro-N-methylbenzylamine, 95%
2-Fluoro-N-methylbenzylamine, 95%
Thermo Scientific Chemicals

2-Fluoro-N-methylbenzylamine, 95%

CAS: 399-30-4 | C8H10FN | 139.173 g/mol
Quantity:
1 g
5 g
Catalog number H54439.03
also known as H54439-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS399-30-4
Chemical Name or Material2-Fluoro-N-methylbenzylamine
ColorColorless to Tan
Density1.047 g/mL
Flash Point68°C (154°F)
View more
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and air. Store at 4-2°C.
RUO – Research Use Only
Natalia Guzior.; Marek Bajda.; Jurand Rakoczy.; Boris Brus.; Stanislav Gobec.; Barbara Malawska. soindoline-1,3-dione derivatives targeting cholinesterases: Design, synthesis and biological evaluation of potential anti-Alzheimer’s agents. Bioorg. Med. Chem. 2015, 23 (7),1629-1637.