Pyrimidine-2-carboxylic acid, 97%
Pyrimidine-2-carboxylic acid, 97%
Pyrimidine-2-carboxylic acid, 97%
Pyrimidine-2-carboxylic acid, 97%
Thermo Scientific Chemicals

Pyrimidine-2-carboxylic acid, 97%

CAS: 31519-62-7 | C5H4N2O2 | 124.10 g/mol
Quantity:
1 g
5 g
Catalog number H54745.06
also known as H54745-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Specifications
CAS31519-62-7
Chemical Name or MaterialPyrimidine-2-carboxylic acid
Formula Weight124.1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
2-carboxy-Pyrimidine is a synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-carboxy-Pyrimidine is a synthetic intermediate.

Solubility
Soluble in ethanol, Dimethyl sulfoxide (DMSO), Dimethylformamide (DMF), and solution of Phosphate Buffered Saline (PBS).

Notes
Store at -20°C.
RUO – Research Use Only

General References:

  1. W. Roger Tully.; Colin R. Gardner.; Roger J. Gillespie.; Robert Westwood. 2-(Oxadiazolyl)- and 2-(thiazolyl)imidazo[1,2-a]pyrimidines as agonists and inverse agonists at benzodiazepine receptors.J. Med. Chem. 1991, 34 (7), 2060-2067.
  2. T. H. Althuis, P. F. Moore, H. J. Hess. Development of ethyl 3,4-dihydro-4-oxopyrimido[4,5-b]quinoline-2-carboxylate, a new prototype with oral antiallergy activity.J. Med. Chem. 1979, 22 (1), 44-48.