2-Amino-4-(4-cyanophenyl)thiazole, 97%
2-Amino-4-(4-cyanophenyl)thiazole, 97%
2-Amino-4-(4-cyanophenyl)thiazole, 97%
Thermo Scientific Chemicals

2-Amino-4-(4-cyanophenyl)thiazole, 97%

CAS: 436151-85-8 | C10H7N3S | 201.25 g/mol
Quantity:
250 mg
1 g
5 g
Catalog number H54861.MD
also known as H54861-MD
Price (JPY)
-
Quantity:
250 mg
Request bulk or custom format
Specifications
Beilstein15029305
CAS436151-85-8
Chemical Name or Material2-Amino-4-(4-cyanophenyl)thiazole
Formula Weight201.25
Health Hazard 1H302-H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only