3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%
3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%
3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%
Thermo Scientific Chemicals

3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%

CAS: 3065-79-0 | C22H24IN2S2+ | 507.472 g/mol
Quantity:
250 mg
1 g
Catalog number H55144.03
also known as H55144-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS3065-79-0
Chemical Name or Material3,3'-Diethyl-9-methylthiacarbocyanine iodide
EINECS Number221-317-2
Formula Weight506.46
Health Hazard 1Warning
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only