N-Boc-3-cyclohexyl-L-alanine, 98%
N-Boc-3-cyclohexyl-L-alanine, 98%
N-Boc-3-cyclohexyl-L-alanine, 98%
N-Boc-3-cyclohexyl-L-alanine, 98%
Thermo Scientific Chemicals

N-Boc-3-cyclohexyl-L-alanine, 98%

CAS: 37736-82-6 | C14H25NO4 | 271.357 g/mol
Quantity:
1 g
5 g
Catalog number H55282.03
also known as H55282-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein3060603
CAS37736-82-6
Chemical Name or MaterialN-Boc-3-cyclohexyl-L-alanine
Formula Weight271.36
IUPAC Name(2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
View more
It is an agrochemical, organic and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
RUO – Research Use Only