Agmatine sulfate, 97%, Thermo Scientific Chemicals
Agmatine sulfate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Agmatine sulfate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H55363.06
also known as H55363-06
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
220.00 
Save 32.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
220.00 
Save 32.35 (15%)
Add to cart
Agmatine sulfate, 97%, Thermo Scientific Chemicals
Catalog numberH55363.06
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
220.00 
Save 32.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameN''-(4-aminobutyl)guanidine; sulfuric acid
Molecular FormulaC5H16N4O4S
InChI KeyPTAYFGHRDOMJGC-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NCCCCN=C(N)N
Molecular Weight (g/mol)228.27
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0% [US]
CommentMaterial sourced in either [US] or [UK]
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% [UK]
FormPowder
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

Solubility
Soluble in water, nearly insoluble in alcohol

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Yoko Sakakibara.; Hiroshi Yanagisawa. Agmatine deiminase from cucumber seedlings is a mono-specific enzyme: purification and characteristics. Protein Expression and Purification. 2003, 30, (1), 88-93.
  2. Kazuho Abe.; Yuzuru Abe.; Hiroshi Saito. Agmatine suppresses nitric oxide production in microglia. Brain Research. 2000, 872, (1-2), 141-148.