L-beta-Proline, 98+%
L-beta-Proline, 98+%
L-beta-Proline, 98+%
Thermo Scientific Chemicals

L-beta-Proline, 98+%

CAS: 72580-53-1 | C5H9NO2 | 115.132 g/mol
Quantity:
250 mg
1 g
Catalog number H57895.03
also known as H57895-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein3537293
CAS72580-53-1
Chemical Name or MaterialL-beta-Proline
Formula Weight115.13
IUPAC Name(3S)-pyrrolidine-3-carboxylic acid
View more
It is an agrochemical and pharmacetical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical and pharmacetical intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents, acids.
RUO – Research Use Only
David Alker.; Kevin J. Doyle.; Laurence M. Harwood.; Andrew McGregor. The direct synthesis of the cyclic sulphamidate of (S)-prolinol: SimultaneousN-protection and activation towards nucleophilic displacement of oxygen. Tetrahedron: Asymmetry. 1990, 1 (12),877-880.