Specifications
CAS1628442-55-6
Chemical Name or Material4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
Formula Weight288.07
IUPAC Name2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
InChI KeyLLJRSXMGPKHUQX-UHFFFAOYSA-N
View more