Azetidine hydrochloride, 97%
Azetidine hydrochloride, 97%
Azetidine hydrochloride, 97%
Thermo Scientific Chemicals

Azetidine hydrochloride, 97%

CAS: 36520-39-5 | C3H8ClN | 93.554 g/mol
Quantity:
1 g
5 g
Catalog number H59935.06
also known as H59935-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS36520-39-5
IUPAC Nameazetidine hydrochloride
Molecular FormulaC3H8ClN
InChI KeyHGQULGDOROIPJN-UHFFFAOYSA-N
SMILESCl.C1CNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to orange to brown
FormPowder or crystalline powder or crystals
Assay from Suppliers CofA≥96.0% (NMR)
Proton NMRConforms to structure
Azetidine hydrochloride is a building block for azetidine synthesis and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine hydrochloride is a building block for azetidine synthesis and a pharmaceutical intermediate.

Solubility
Soluble in water (103.8°C at 760 mmHg).

Notes
Hygroscopic. Reacts with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Christopher S. Dunkley.; Charles J. Thoman. Synthesis and biological evaluation of a novel phenyl substituted sydnone series as potential antitumor agents. Bioorg. Med. Chem. Lett. 2003, 13 (17), 2899-2901.
  2. Paritosh R. Dave. Acylative Dealkylation of N-tert-Butyl-3-substituted Azetidines: Facile Access to [1.1.0]Azabicyclobutane, 3-Hydroxyazetidinium Hydrochloride, and 3-Azetidinones. J. Org. Chem. 1996, 61 (6),5453-5455.