Diethyl iminodiacetate, 97%
Diethyl iminodiacetate, 97%
Thermo Scientific Chemicals

Diethyl iminodiacetate, 97%

CAS: 6290-05-7 | C8H15NO4 | 189.21 g/mol
Quantity:
5 mL
25 mL
Catalog number H60608.AA
also known as H60608-AA
Price (JPY)
-
Request A Quote
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
CAS6290-05-7
IUPAC Nameethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Molecular FormulaC8H15NO4
InChI KeyLJDNMOCAQVXVKY-UHFFFAOYSA-N
SMILESCCOC(=O)CNCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

Solubility
Moderately soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Shiu-Chun Chuang; Chin-Yuan Chang; Chuen-Ying Liu. Polystyrene monolithic column functionalized with copper-iminodiacetate complex as a stationary phase for open tubular capillary electrochromatography. Journal of Chromatography A . 2004, 1044 (1-2), 229-236.
  2. M Colvin; R B Brundrett; M N Kan; I Jardine; C Fenselau. Alkylating properties of phosphoramide mustard. Cancer Research. 1976, 36 (3), 1121-1126.