4-Hydroxy-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals
4-Hydroxy-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals
4-Hydroxy-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals
4-Hydroxy-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog NumberQuantity
H60810.09
also known as H60810-09
10 g
Catalog number H60810.09
also known as H60810-09
Price (JPY)
-
Request A Quote
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS578-39-2
IUPAC Name4-hydroxy-2-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyBBMFSGOFUHEVNP-UHFFFAOYSA-N
SMILESCC1=CC(O)=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only